This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF2 SIRNA Assays ›
Gene Symbol:
Rnf2
Gene Name:
ring finger protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 68829714 - 68859929 on Build Rnor_6.0
UniGene ID:
Rn.19719
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS315271
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025667.1 NP_001020838.1 3 342
XM_006249990.2 XP_006250052.1 3 280
XM_006249991.2 XP_006250053.1 3 362
GenBank mRNA AB568264.1 - 2 146
BC098941.1 - 3 342 7456372

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
gastrulation with mouth forming second
transcription, DNA-templated
anterior/posterior axis specification
histone ubiquitination
spinal cord development
response to retinoic acid
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
molecular_function
chromatin binding
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity
RING-like zinc finger domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0