This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC93 SIRNA Assays ›
Gene Symbol:
Ccdc93
Gene Name:
coiled-coil domain containing 93
Gene Aliases:
-
Chromosome Location:
Chr.13: 37400460 - 37473973 on Build Rnor_6.0
UniGene ID:
Rn.79004
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS315197
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc93
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024997.1 NP_001020168.1 20, 21 1744
XM_008769456.1 XP_008767678.1 21, 22 1778
GenBank mRNA BC091337.1 - 20, 21 1744 7314934

Back To Top

More Information


Gene Ontology Categories:

Function(s)

Golgi to plasma membrane transport
protein transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0