This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MUC20 SIRNA Assays ›
Gene Symbol:
Muc20
Gene Name:
mucin 20, cell surface associated
Gene Aliases:
-
Chromosome Location:
Chr.11: 71222605 - 71236223 on Build Rnor_6.0
UniGene ID:
Rn.93766
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS314675
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Muc20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107089.2 NP_001100559.1 2 1002
XM_006248449.2 XP_006248511.1 2 1771
GenBank mRNA BC167011.1 - 2 1002 7128599

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
biological_process
hepatocyte growth factor receptor signaling pathway
protein homooligomerization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm