siRNA Details
Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012005.2 | NP_001012005.2 | 20 | 4173 | ||
GenBank mRNA | BC088325.1 | - | 4 | 771 | ||
BC169012.1 | - | 20 | 4173 | 9022732 |

Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012005.2 | NP_001012005.2 | 20 | 4173 | ||
GenBank mRNA | BC088325.1 | - | 4 | 771 | ||
BC169012.1 | - | 20 | 4173 | 9022732 |