siRNA Details
Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012005.2 | NP_001012005.2 | 7 | 2046 | ||
GenBank mRNA | BC169012.1 | - | 7 | 2046 | 9022732 |

Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012005.2 | NP_001012005.2 | 7 | 2046 | ||
GenBank mRNA | BC169012.1 | - | 7 | 2046 | 9022732 |