This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP39A1 SIRNA Assays ›
Gene Symbol:
Cyp39a1
Gene Name:
cytochrome P450, family 39, subfamily a, polypeptide 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 19804079 - 19880346 on Build Rnor_6.0
UniGene ID:
Rn.153360
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS313254
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp39a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106893.1 NP_001100363.1 6 1115
XM_006244604.2 XP_006244666.1 6 1115
XM_006244605.1 XP_006244667.1 6 1115
XM_008766854.1 XP_008765076.1 6 998
XM_008766855.1 XP_008765077.1 6 996

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
cholesterol catabolic process
biological_process
oxidation-reduction process
molecular_function
iron ion binding
steroid 7-alpha-hydroxylase activity
oxysterol 7-alpha-hydroxylase activity
heme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0