This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
Lrrfip2
Gene Name:
leucine rich repeat (in FLII) interacting protein 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 119381666 - 119484125 on Build Rnor_6.0
UniGene ID:
Rn.20528
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS313107
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrfip2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024761.1 NP_001019932.1 16 1559
XM_006243939.2 XP_006244001.1 28 2344
XM_006243957.1 XP_006244019.1 15 1598
XM_006243958.2 XP_006244020.1 15 1538
XM_006243960.1 XP_006244022.1 14 1526
XM_008766616.1 XP_008764838.1 30 2540
XM_008766617.1 XP_008764839.1 30 2303
XM_008766618.1 XP_008764840.1 30 2516
XM_008766619.1 XP_008764841.1 29 2469
XM_008766620.1 XP_008764842.1 29 2495
XM_008766621.1 XP_008764843.1 29 2489
XM_008766622.1 XP_008764844.1 29 2471
XM_008766623.1 XP_008764845.1 29 2470
XM_008766624.1 XP_008764846.1 29 2450
XM_008766625.1 XP_008764847.1 29 2438
XM_008766626.1 XP_008764848.1 28 2420
XM_008766627.1 XP_008764849.1 28 2380
XM_008766628.1 XP_008764850.1 27 2374
XM_008766629.1 XP_008764851.1 26 2305
GenBank mRNA BC097955.1 - 16 1559 7456701
FQ233539.1 - 15 1582

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Wnt signaling pathway
LRR domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0