This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC26A6 SIRNA Assays ›
Gene Symbol:
Slc26a6
Gene Name:
solute carrier family 26 (anion exchanger), member 6
Gene Aliases:
-
Chromosome Location:
Chr.8: 117648796 - 117659376 on Build Rnor_6.0
UniGene ID:
Rn.18851
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS313082
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc26a6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143817.1 NP_001137289.1 9 1051
XM_006243755.2 XP_006243817.1 9 3079
XM_006243756.2 XP_006243818.1 9 2257
XM_006243757.2 XP_006243819.1 9 1623
XM_006243758.2 XP_006243820.1 9 1624
XM_008766510.1 XP_008764732.1 8 1519

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chloride transport
sulfate transport
bicarbonate transport
formate transport
mannitol transport
oxalate transport
transepithelial chloride transport
angiotensin-activated signaling pathway
epithelial fluid transport
oxalic acid secretion
sperm capacitation
intestinal absorption
regulation of intracellular pH
intracellular pH elevation
protein kinase C signaling
transepithelial transport
cellular response to cAMP
cellular response to fructose stimulus
cellular response to interferon-gamma
anion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
positive regulation of dipeptide transmembrane transport
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
formate transmembrane transporter activity
efflux transmembrane transporter activity
formate uptake transmembrane transporter activity
formate efflux transmembrane transporter activity
oxalate transmembrane transporter activity
PDZ domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0