This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NCKIPSD SIRNA Assays ›
Gene Symbol:
Nckipsd
Gene Name:
NCK interacting protein with SH3 domain
Gene Aliases:
-
Chromosome Location:
Chr.8: 117601027 - 117612302 on Build Rnor_6.0
UniGene ID:
Rn.90103
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS313078
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nckipsd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106857.1 NP_001100327.1 12 1969
XM_006243753.2 XP_006243815.1 12 1982
XM_006243754.2 XP_006243816.1 12 2327
XM_008766508.1 XP_008764730.1 12 4095
XM_008766509.1 XP_008764731.1 13 2121

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0