This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMOD3 SIRNA Assays ›
Gene Symbol:
Tmod3
Gene Name:
tropomodulin 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 82432165 - 82492243 on Build Rnor_6.0
UniGene ID:
Rn.146101
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS312975
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmod3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011997.1 NP_001011997.1 6 633
GenBank mRNA BC079290.1 - 6 633 7134366
FQ229312.1 - 1 67

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016963, AK169218, AK084463, AF237630, BC082595, and others.

Gene Ontology Categories:

Function(s) Process(es)

muscle contraction
actin filament organization
actin cytoskeleton organization
myofibril assembly
erythrocyte development
pointed-end actin filament capping
positive regulation of mitotic cell cycle phase transition
tropomyosin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0