This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FOXH1 SIRNA Assays ›
Gene Symbol:
Foxh1
Gene Name:
forkhead box H1
Gene Aliases:
-
Chromosome Location:
Chr.7: 117730307 - 117732666 on Build Rnor_6.0
UniGene ID:
Rn.100970
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS312532
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Foxh1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130493.1 NP_001123965.1 1 185
XM_008765565.1 XP_008763787.1 1 464
GenBank mRNA BC158731.1 - 1 137

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007989, BC137781, AF069303, AF110506, AF079514, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
heart looping
secondary heart field specification
outflow tract morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transforming growth factor beta receptor signaling pathway
determination of left/right symmetry
anterior/posterior pattern specification
negative regulation of intracellular estrogen receptor signaling pathway
embryonic heart tube anterior/posterior pattern specification
aorta morphogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
axial mesoderm development
negative regulation of androgen receptor signaling pathway
negative regulation of androgen receptor activity
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription cofactor activity
transcriptional activator activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein domain specific binding
bHLH transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
SMAD binding
androgen receptor binding
co-SMAD binding
R-SMAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0