This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB3IP SIRNA Assays ›
Gene Symbol:
Rab3ip
Gene Name:
RAB3A interacting protein
Gene Aliases:
RABIN3
Chromosome Location:
Chr.7: 59927486 - 59971516 on Build Rnor_6.0
UniGene ID:
Rn.31889
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS311929
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab3ip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017313.3 NP_059009.2 1 254
XM_006241357.2 XP_006241419.1 2 423
XM_006241358.2 XP_006241420.1 2 428
XM_006241359.2 XP_006241421.1 2 1432
XM_006241360.2 XP_006241422.1 2 418
XM_008765382.1 XP_008763604.1 1 480
GenBank mRNA BC127500.1 - 1 257 7457420
U19181.1 - 1 247

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
protein transport
protein localization to organelle
cilium assembly
protein binding
Rab guanyl-nucleotide exchange factor activity
GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0