This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100294508 SIRNA Assays ›
Gene Symbol:
LOC100294508
Gene Name:
dyslexia susceptibility 2-like
Gene Aliases:
-
Chromosome Location:
Chr.5: 144779391 - 144872271 on Build Rnor_6.0
UniGene ID:
Rn.229753
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS311759
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100294508
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159655.3 NP_001153127.1 7 1338
XM_006238841.2 XP_006238903.1 7 1293
XM_006238842.2 XP_006238904.1 7 1267
XM_008763984.1 XP_008762206.1 7 1710
GenBank mRNA CO567907.1 - 4 348

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0