siRNA Details
Gene Symbol: Ttc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106671.1 | NP_001100141.1 | 7 | 1478 | ||
XM_006238502.2 | XP_006238564.1 | 4 | 2030 |

Gene Symbol: Ttc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106671.1 | NP_001100141.1 | 7 | 1478 | ||
XM_006238502.2 | XP_006238564.1 | 4 | 2030 |