This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEMA6C SIRNA Assays ›
Gene Symbol:
Sema6c
Gene Name:
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C
Gene Aliases:
-
Chromosome Location:
Chr.2: 196330795 - 196347218 on Build Rnor_6.0
UniGene ID:
Rn.22449
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS311168
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sema6c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017308.1 NP_059004.1 11 1172
XM_006232858.2 XP_006232920.1 14 1584
XM_006232861.2 XP_006232923.1 12 1446
XM_006232869.2 XP_006232931.1 10 1093
XM_008761286.1 XP_008759508.1 12 4850
XM_008761287.1 XP_008759509.1 12 4197
XM_008761288.1 XP_008759510.1 12 3123
XM_008761289.1 XP_008759511.1 12 4702
XM_008761290.1 XP_008759512.1 12 3949
XM_008761291.1 XP_008759513.1 12 4280
XM_008761292.1 XP_008759514.1 12 3379
XM_008761293.1 XP_008759515.1 13 1536
XM_008761294.1 XP_008759516.1 11 5008
GenBank mRNA AB000817.2 - 11 1188
AB014074.1 - 11 1188
AF363971.1 - 10 1052

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

axon guidance
receptor activity
semaphorin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0