This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COPG1 SIRNA Assays ›
Gene Symbol:
Copg1
Gene Name:
coatomer protein complex, subunit gamma 1
Gene Aliases:
Copg
Chromosome Location:
Chr.4: 119815041 - 119839683 on Build Rnor_6.0
UniGene ID:
Rn.50150
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS311153
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Copg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031822.3 NP_001026992.1 23 2440
XM_008763066.1 XP_008761288.1 23 2645
GenBank mRNA AB158425.1 - 23 2440
AB158426.1 - 23 2440
BC129074.1 - 23 2531 7107382

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle-mediated transport
establishment of Golgi localization
organelle transport along microtubule
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0