This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF2AK3 SIRNA Assays ›
Gene Symbol:
Eif2ak3
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases:
PEK
Chromosome Location:
Chr.4: 98648507 - 98709695 on Build Rnor_6.0
UniGene ID:
Rn.24897
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS311039
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif2ak3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031599.2 NP_113787.1 9 1751
XM_006236623.1 XP_006236685.1 9 1269
XM_006236624.2 XP_006236686.1 9 1789
XM_008762977.1 XP_008761199.1 8 1383
GenBank mRNA AF096835.1 - 9 1752

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
ossification
angiogenesis
chondrocyte development
translation
protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
ER overload response
endoplasmic reticulum organization
lactation
positive regulation of signal transduction
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of gene expression
regulation of translational initiation by eIF2 alpha phosphorylation
negative regulation of translation
regulation of fatty acid metabolic process
calcium-mediated signaling
bone mineralization
endoplasmic reticulum unfolded protein response
pancreas development
endocrine pancreas development
negative regulation of myelination
negative regulation of translation in response to stress
negative regulation of translational initiation in response to stress
positive regulation of protein binding
SREBP signaling pathway
response to endoplasmic reticulum stress
cellular response to glucose starvation
negative regulation of apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase I promoter
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
protein homooligomerization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding
enzyme binding
protein phosphatase binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0