This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMA5 SIRNA Assays ›
Gene Symbol:
Psma5
Gene Name:
proteasome (prosome, macropain) subunit, alpha type 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 211050385 - 211073703 on Build Rnor_6.0
UniGene ID:
Rn.1276
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310889
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psma5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017282.1 NP_058978.1 7 574
GenBank mRNA BC060575.1 - 7 595 6920479
D10756.1 - 7 574
FQ218317.1 - 7 591

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
threonine-type endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header