This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FUT7 SIRNA Assays ›
Gene Symbol:
Fut7
Gene Name:
fucosyltransferase 7 (alpha (1,3) fucosyltransferase)
Gene Aliases:
-
Chromosome Location:
Chr.3: 2637385 - 2647738 on Build Rnor_6.0
UniGene ID:
Rn.139741
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310762
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fut7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199491.1 NP_955785.1 3 1492
XM_006233585.2 XP_006233647.1 2 1378
XM_006233586.2 XP_006233648.1 2 1072
XM_006233587.2 XP_006233649.1 2 1101
XM_008761582.1 XP_008759804.1 2 1176
XM_008761583.1 XP_008759805.1 2 1327
GenBank mRNA AJ276068.2 - 3 1592

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
leukocyte migration involved in immune response
protein glycosylation
fucosylation
fucosyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0