This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANAPC2 SIRNA Assays ›
Gene Symbol:
Anapc2
Gene Name:
anaphase promoting complex subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 2490470 - 2502247 on Build Rnor_6.0
UniGene ID:
Rn.20099
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310757
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anapc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100532.1 NP_001094002.1 10 1800
GenBank mRNA BC107471.1 - 8 904
BC166796.1 - 10 1800 7938000
FQ224461.1 - 10 1810

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175300, AA727217, BC027351, AK129351, AK191347, and others.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
positive regulation of synaptic plasticity
positive regulation of axon extension
positive regulation of dendrite morphogenesis
protein K11-linked ubiquitination
positive regulation of synapse maturation
molecular_function
ubiquitin protein ligase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header