This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZGPAT SIRNA Assays ›
Gene Symbol:
Zgpat
Gene Name:
zinc finger, CCCH-type with G patch domain
Gene Aliases:
RGD1310801
Chromosome Location:
Chr.3: 176865113 - 176881603 on Build Rnor_6.0
UniGene ID:
Rn.8219
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310722
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zgpat
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009656.1 NP_001009656.1 2 721
XM_006235749.1 XP_006235811.1 3 717
XM_006235750.1 XP_006235812.1 3 682
XM_006235751.2 XP_006235813.1 3 703
XM_006235752.2 XP_006235814.1 2 617
GenBank mRNA BC087720.1 - 2 721 7318724

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0