This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGAT3 SIRNA Assays ›
Gene Symbol:
Mgat3
Gene Name:
mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.7: 121394541 - 121436935 on Build Rnor_6.0
UniGene ID:
Rn.9803
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310239
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mgat3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019239.2 NP_062112.2 2 1105
XM_006242072.2 XP_006242134.1 2 1200
XM_006242073.2 XP_006242135.1 2 1675
XM_006242074.2 XP_006242136.1 2 1078
XM_006242077.2 XP_006242139.1 2 1564
GenBank mRNA D10852.1 - 2 1105

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
beta-1,4-mannosylglycoprotein 4-beta-N-acetylglucosaminyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0