This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SESTD1 SIRNA Assays ›
Gene Symbol:
Sestd1
Gene Name:
SEC14 and spectrin domains 1
Gene Aliases:
RGD1562244
Chromosome Location:
Chr.3: 64044111 - 64134977 on Build Rnor_6.0
UniGene ID:
Rn.18006
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS310121
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sestd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134514.2 NP_001127986.2 16 1847
XM_006234402.2 XP_006234464.1 17 2127
XM_006234404.2 XP_006234466.1 16 2037
XM_008761892.1 XP_008760114.1 17 2053
XM_008761893.1 XP_008760115.1 16 1963
GenBank mRNA BC169110.1 - 16 1847 7445081

Back To Top

More Information


Gene Ontology Categories:

Process(es)

phosphatidylserine binding
1-phosphatidylinositol binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-5-phosphate binding
phosphatidylcholine binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0