This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AMHR2 SIRNA Assays ›
Gene Symbol:
Amhr2
Gene Name:
anti-Mullerian hormone receptor, type II
Gene Aliases:
-
Chromosome Location:
Chr.7: 144052202 - 144060678 on Build Rnor_6.0
UniGene ID:
Rn.10165
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS309839
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Amhr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030998.1 NP_112260.1 11 1703
GenBank mRNA KJ923229.1 - 10 1566
U42427.1 - 11 1703
X71916.1 - 11 1685

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144547, BC119572, BC119571, AF503863.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
transforming growth factor beta receptor signaling pathway
G-protein coupled receptor signaling pathway
sex differentiation
signal transduction by protein phosphorylation
negative regulation of anti-Mullerian hormone signaling pathway
anti-Mullerian hormone signaling pathway
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
receptor activity
transforming growth factor beta receptor activity, type II
ATP binding
hormone binding
metal ion binding
anti-Mullerian hormone receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0