This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC102551660 SIRNA Assays ›
Gene Symbol:
LOC102551660
Gene Name:
echinoderm microtubule-associated protein-like 3-like
Gene Aliases:
-
Chromosome Location:
Chr.1: 225129008 - 225140472 on Build Rnor_6.0
UniGene ID:
Rn.11674
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS308696
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC102551660
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231007.2 XP_006231069.1 7, 8 1200
XM_006231008.1 XP_006231070.1 7, 8 1170
XM_006231009.2 XP_006231071.1 7, 8 1201
GenBank mRNA FQ228620.1 - 7, 8 1170

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_153265, BC108280, AK093146, AL833388, BC075824, and others.
This siRNA also targets the following transcripts in Mouse: NM_144872, BC023878, BC022146.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0