This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SIRT3 SIRNA Assays ›
Gene Symbol:
Sirt3
Gene Name:
sirtuin 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 213613502 - 213636194 on Build Rnor_6.0
UniGene ID:
Rn.24698
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS308592
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sirt3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106313.2 NP_001099783.1 3 286
XM_006230526.2 XP_006230588.1 3 366
XM_006230529.2 XP_006230591.1 3 980
XM_008759979.1 XP_008758201.1 3 406
XM_008759980.1 XP_008758202.1 3 406
XM_008759981.1 XP_008758203.1 3 274
GenBank mRNA BC167006.1 - 3 286 7314283
EU886468.1 - 3 312
FQ213357.1 - 2 112
FQ218084.1 - 3 317

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein deacetylation
aging
aerobic respiration
positive regulation of insulin secretion
peptidyl-lysine deacetylation
negative regulation of ERK1 and ERK2 cascade
histone H3 deacetylation
negative regulation of reactive oxygen species metabolic process
negative regulation of peptidyl-lysine acetylation
zinc ion binding
enzyme binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
NAD+ binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header