This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC89 SIRNA Assays ›
Gene Symbol:
Ccdc89 LOC100911776
Gene Name:
coiled-coil domain containing 89
coiled-coil domain-containing protein 89-like
Gene Aliases:
RGD1305335
-
Chromosome Location:
Chr.1: 154606461 - 154607723 on Build Rnor_6.0
Chromosome Location:
Chr.1: 156325241 - 156328892 on Build Rnor_6.0
UniGene ID:
Rn.108932
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS308130
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc89
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134833.2 NP_001128305.1 1 1019
GenBank mRNA BC167063.1 - 1 1019 7453194
Gene Symbol: LOC100911776
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748922.3 XP_003748970.1 1 2559

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0