This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAP4K1 SIRNA Assays ›
Gene Symbol:
Map4k1
Gene Name:
mitogen activated protein kinase kinase kinase kinase 1
Gene Aliases:
Hpk1
Chromosome Location:
Chr.1: 87937123 - 87959514 on Build Rnor_6.0
UniGene ID:
Rn.137063
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS307928
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Map4k1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106243.1 NP_001099713.1 32 2573
XM_006228716.2 XP_006228778.1 32 3532
XM_008759122.1 XP_008757344.1 30 3397

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001042600, NM_007181, AW137524, U66464.
This siRNA also targets the following transcripts in Mouse: NM_008279, BC005433, AK143215, AK011024, BC024832, and others.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPKKK activity
protein phosphorylation
biological_process
cell proliferation
peptidyl-serine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
protein autophosphorylation
molecular_function
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
MAP kinase kinase kinase kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0