This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TDRD1 SIRNA Assays ›
Gene Symbol:
Tdrd1
Gene Name:
tudor domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 277637474 - 277685236 on Build Rnor_6.0
UniGene ID:
Rn.156064
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS307569
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdrd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106202.1 NP_001099672.1 20 2998
XM_006231642.2 XP_006231704.1 21 3261
XM_008760524.1 XP_008758746.1 21 3068
XM_008760525.1 XP_008758747.1 21 3033
XM_008760526.1 XP_008758748.1 20 3198
XM_008760528.1 XP_008758750.1 20 2827
XM_008760529.1 XP_008758751.1 20 3105
XM_008760530.1 XP_008758752.1 20 3018
XM_008760531.1 XP_008758753.1 19 2847

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

germ cell development
spermatogenesis
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header