This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRAPPC8 SIRNA Assays ›
Gene Symbol:
Trappc8 LOC103689930
Gene Name:
trafficking protein particle complex 8
trafficking protein particle complex subunit 8
Gene Aliases:
RGD1309102
-
Chromosome Location:
Chr.18: 12070081 - 12116697 on Build Rnor_6.0
Chromosome Location:
Chr.18: 15298678 - 15375034 on Build Rnor_6.0
UniGene ID:
Rn.6435
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS307266
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trappc8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106160.1 NP_001099630.1 2851
GenBank mRNA FQ230517.1 - 708
Gene Symbol: LOC103689930
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006254461.2 XP_006254523.1 20 3097
XM_006254462.2 XP_006254524.1 20 3094
XM_008771994.1 XP_008770216.1 20 3151

Back To Top

More Information


Gene Ontology Categories:

Function(s)

autophagosome assembly
ER to Golgi vesicle-mediated transport
pexophagy
CVT pathway
protein localization to pre-autophagosomal structure
nucleophagy

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm