This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100911363 SIRNA Assays ›
Gene Symbol:
LOC100911363 Capns1 LOC100912380
Gene Name:
calpain small subunit 1-like
calpain, small subunit 1
calpain small subunit 1-like
Gene Aliases:
-
Capn4
-
Chromosome Location:
Chr.1: 91058368 - 91064929 on Build Rnor_6.0
Chromosome Location:
Chr.1: 90918124 - 91071025 on Build Rnor_6.0
Chromosome Location:
Chr.1: 88652448 - 88656835 on Build Rnor_6.0
UniGene ID:
-
Rn.3430
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID RSS307153
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100911363
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748836.3 XP_003748884.1 6 550
XM_008759263.1 XP_008757485.1 6 752
Gene Symbol: Capns1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017118.1 NP_058814.1 6 501
GenBank mRNA BC098068.1 - 6 501 7462537
FQ229527.1 - 184
U10861.1 - 3 190
U53859.1 - 5 437
Gene Symbol: LOC100912380
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228796.2 XP_006228858.2 5 277

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
calcium ion binding
calcium-dependent cysteine-type endopeptidase activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header