This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other JAG1 SIRNA Assays ›
Gene Symbol:
Jag1
Gene Name:
jagged 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 130079361 - 130114781 on Build Rnor_6.0
UniGene ID:
Rn.88804
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS307097
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Jag1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019147.1 NP_062020.1 8 1459
GenBank mRNA L38483.1 - 8 1459

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell fate determination
blood vessel remodeling
morphogenesis of an epithelial sheet
T cell mediated immunity
pulmonary valve morphogenesis
cardiac right ventricle morphogenesis
Notch signaling pathway
organ morphogenesis
organ regeneration
response to muramyl dipeptide
aorta morphogenesis
regulation of cell proliferation
auditory receptor cell differentiation
camera-type eye development
negative regulation of cell differentiation
negative regulation of fat cell differentiation
positive regulation of myeloid cell differentiation
negative regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
inner ear development
cardiac septum morphogenesis
ciliary body morphogenesis
pulmonary artery morphogenesis
cardiac neural crest cell development involved in outflow tract morphogenesis
Notch signaling involved in heart development
endocardial cushion cell development
distal tubule development
loop of Henle development
neuronal stem cell population maintenance
negative regulation of stem cell differentiation
Notch binding
calcium ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0