This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CANX SIRNA Assays ›
Gene Symbol:
Canx
Gene Name:
calnexin
Gene Aliases:
-
Chromosome Location:
Chr.10: 35800942 - 35833937 on Build Rnor_6.0
UniGene ID:
Rn.1762
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS307088
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Canx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172008.2 NP_742005.1 12, 13 1613
XM_008767679.1 XP_008765901.1 12, 13 1734
XM_008767680.1 XP_008765902.1 12, 13 1653
GenBank mRNA FQ210638.1 - 12, 13 1639
L18889.1 - 11, 12 1510

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001024649, NM_001746, AK130080, L18887, M98452, and others.
This siRNA also targets the following transcripts in Mouse: NM_001110499, NM_007597, NM_001110500, BC040244, L18888, and others.

Gene Ontology Categories:

Function(s) Process(es)

aging
synaptic vesicle endocytosis
chaperone-mediated protein folding
clathrin-mediated endocytosis
glycoprotein binding
calcium ion binding
protein binding
carbohydrate binding
apolipoprotein binding
ionotropic glutamate receptor binding
poly(A) RNA binding
unfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header