This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAF2 SIRNA Assays ›
Gene Symbol:
Faf2
Gene Name:
Fas associated factor family member 2
Gene Aliases:
2210404d11rik, RGD1306577, Ubxd8
Chromosome Location:
Chr.17: 10485650 - 10527882 on Build Rnor_6.0
UniGene ID:
Rn.111214
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS306849
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Faf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017445.1 NP_001017445.1 9 811
XM_006253617.2 XP_006253679.1 8 836
XM_008771488.1 XP_008769710.1 6 620
GenBank mRNA BC079356.1 - 8 810
BC091224.1 - 9 811 7128668

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to unfolded protein
lipid particle organization
negative regulation of catalytic activity
ubiquitin protein ligase binding
lipase binding
ubiquitin binding
lipase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0