This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CATSPER3 SIRNA Assays ›
Gene Symbol:
Catsper3
Gene Name:
cation channel, sperm associated 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 8900034 - 8926546 on Build Rnor_6.0
UniGene ID:
Rn.187544
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS306843
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Catsper3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106101.1 NP_001099571.1 2 233
XM_006253572.2 XP_006253634.1 2 1433

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
sperm motility
regulation of ion transmembrane transport
sperm capacitation
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0