This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IRF2 SIRNA Assays ›
Gene Symbol:
Irf2
Gene Name:
interferon regulatory factor 2
Gene Aliases:
-
Chromosome Location:
Chr.16: 48582354 - 48732129 on Build Rnor_6.0
UniGene ID:
Rn.107887
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS306714
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Irf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047086.1 NP_001040551.1 4 399
XM_006253139.2 XP_006253201.1 4 660
XM_008771256.1 XP_008769478.1 4 495
GenBank mRNA BC166593.1 - 4 440 7934031
DQ674266.1 - 4 399

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008391, AK155025, AK087926, BC006577, AK158642, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulatory region DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0