This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other USP46 SIRNA Assays ›
Gene Symbol:
Usp46
Gene Name:
ubiquitin specific peptidase 46
Gene Aliases:
RGD1564808
Chromosome Location:
Chr.14: 36687134 - 36755360 on Build Rnor_6.0
UniGene ID:
Rn.233806
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS306194
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp46
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191596.1 NP_001178525.1 1 95
GenBank mRNA FQ211578.1 - 1 143
GU455413.1 - 1 19

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_022832, GU455414, AK299883, CN483539, AK296493, and others.
This siRNA also targets the following transcripts in Mouse: NM_177561, AK145298, BC039916.

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
ubiquitin-dependent protein catabolic process
adult feeding behavior
protein deubiquitination
regulation of synaptic transmission, GABAergic
behavioral response to ethanol
righting reflex
thiol-dependent ubiquitin-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header