siRNA Details
Gene Symbol: Vom2r69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099468.1 | NP_001092938.1 | 3 | 1041 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
