This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CR2 SIRNA Assays ›
Gene Symbol:
Cr2
Gene Name:
complement component (3d/Epstein Barr virus) receptor 2
Gene Aliases:
Cd21
Chromosome Location:
Chr.13: 113890274 - 113927814 on Build Rnor_6.0
UniGene ID:
Rn.23490
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS306091
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105989.2 NP_001099459.2 6 941
XM_006250476.2 XP_006250538.1 10 1556
XM_006250477.2 XP_006250539.1 6 1010
XM_008769847.1 XP_008768069.1 13 2141
XM_008769848.1 XP_008768070.1 6 1427

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

complement receptor mediated signaling pathway
B cell differentiation
B cell proliferation
B cell activation
complement binding
DNA binding
receptor activity
complement receptor activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0