This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LMX1A SIRNA Assays ›
Gene Symbol:
Lmx1a
Gene Name:
LIM homeobox transcription factor 1 alpha
Gene Aliases:
-
Chromosome Location:
Chr.13: 85917956 - 86061536 on Build Rnor_6.0
UniGene ID:
Rn.137284
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS305938
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lmx1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105967.2 NP_001099437.2 8 1332
XM_008769728.1 XP_008767950.1 8 1225

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
regulation of transcription, DNA-templated
axon guidance
central nervous system development
brain development
regulation of gene expression
dentate gyrus development
cerebellum development
hippocampus development
central nervous system neuron differentiation
neuron differentiation
midbrain development
negative regulation of neuron differentiation
dopaminergic neuron differentiation
molecular_function
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0