siRNA Details
Gene Symbol: Lmx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105967.2 | NP_001099437.2 | 1 | 235 | ||
XM_008769728.1 | XP_008767950.1 | 1 | 128 |

Gene Symbol: Lmx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105967.2 | NP_001099437.2 | 1 | 235 | ||
XM_008769728.1 | XP_008767950.1 | 1 | 128 |