This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HELZ SIRNA Assays ›
Gene Symbol:
Helz
Gene Name:
helicase with zinc finger
Gene Aliases:
-
Chromosome Location:
Chr.10: 95768709 - 95906992 on Build Rnor_6.0
UniGene ID:
Rn.19942
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS305101
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Helz
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105848.2 NP_001099318.2 13 1854
XM_006247562.2 XP_006247624.1 15 1975
XM_006247565.2 XP_006247627.1 15 1975
XM_008768314.1 XP_008766536.1 16 2026
XM_008768315.1 XP_008766537.1 16 2026
XM_008768316.1 XP_008766538.1 14 1918

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198298, AB093209, BC076626.

Gene Ontology Categories:

Process(es)

poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0