siRNA Details
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105801.2 | NP_001099271.2 | 18 | 3245 | ||
XM_006246665.2 | XP_006246727.1 | 18 | 3602 |

Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105801.2 | NP_001099271.2 | 18 | 3245 | ||
XM_006246665.2 | XP_006246727.1 | 18 | 3602 |