This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALOXE3 SIRNA Assays ›
Gene Symbol:
Aloxe3
Gene Name:
arachidonate lipoxygenase 3
Gene Aliases:
e-LOX-3, eLOX-3
Chromosome Location:
Chr.10: 55711999 - 55735915 on Build Rnor_6.0
UniGene ID:
Rn.8716
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS304839
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aloxe3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105793.1 NP_001099263.1 6 782
XM_006246617.2 XP_006246679.1 7 1414
XM_006246619.2 XP_006246681.1 6 854

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sphingolipid metabolic process
biological_process
epidermis development
sensory perception of pain
arachidonic acid metabolic process
lipoxygenase pathway
peroxisome proliferator activated receptor signaling pathway
linoleic acid metabolic process
fat cell differentiation
ceramide biosynthetic process
hepoxilin biosynthetic process
oxidation-reduction process
establishment of skin barrier
bicellular tight junction assembly
molecular_function
iron ion binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
lyase activity
hepoxilin A3 synthase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0