This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GCNT3 SIRNA Assays ›
Gene Symbol:
Gcnt3
Gene Name:
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases:
beta-16-N-acetylglucosaminyltransferase, dI/C2/C4GnT
Chromosome Location:
Chr.8: 76449078 - 76459434 on Build Rnor_6.0
UniGene ID:
Rn.43518
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS304589
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gcnt3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173312.2 NP_775434.1 2 304
XM_006243366.2 XP_006243428.1 5 999
XM_008766220.1 XP_008764442.1 4 511
GenBank mRNA AB098520.1 - 2 305

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immunoglobulin production in mucosal tissue
protein glycosylation
tissue morphogenesis
intestinal absorption
kidney morphogenesis
beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm