This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HAS3 SIRNA Assays ›
Gene Symbol:
Has3
Gene Name:
hyaluronan synthase 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 39063319 - 39077745 on Build Rnor_6.0
UniGene ID:
Rn.163402
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS304396
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Has3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172319.1 NP_758822.1 4 1497
XM_006255561.2 XP_006255623.1 4 2717
GenBank mRNA AB097569.1 - 4 1497
AF543196.1 - 3 955

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008217, AK028582, U86408, BC138152, BC138161.

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
hyaluronan biosynthetic process
extracellular polysaccharide biosynthetic process
positive regulation of transcription, DNA-templated
extracellular matrix assembly
positive regulation of hyaluranon cable assembly
hyaluronan synthase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0