This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SELP SIRNA Assays ›
Gene Symbol:
Selp
Gene Name:
selectin P
Gene Aliases:
PSELECT
Chromosome Location:
Chr.13: 82428943 - 82464629 on Build Rnor_6.0
UniGene ID:
Rn.10012
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303963
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Selp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013114.1 NP_037246.1 6 1096
XM_006250150.2 XP_006250212.1 7 1210
GenBank mRNA BC101919.1 - 6 1086 7935390
L23088.1 - 6 1096

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte migration
regulation of cellular extravasation
inflammatory response
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
positive regulation of platelet activation
positive regulation of phosphatidylinositol 3-kinase signaling
response to organic cyclic compound
single organismal cell-cell adhesion
response to lipopolysaccharide
regulation of integrin activation
positive regulation of cell adhesion
blood vessel morphogenesis
leukocyte migration
leukocyte tethering or rolling
lipopolysaccharide binding
glycoprotein binding
heparin binding
carbohydrate binding
sialic acid binding
fucose binding
calcium-dependent protein binding
oligosaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0