This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A1 SIRNA Assays ›
Gene Symbol:
Slc7a1
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
Gene Aliases:
Atrc1, CAT1, Cat-1
Chromosome Location:
Chr.12: 8085012 - 8108972 on Build Rnor_6.0
UniGene ID:
Rn.9439
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303958
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013111.3 NP_037243.2 769
XM_008768943.1 XP_008767165.1 2 1041
XM_008768944.1 XP_008767166.1 2 1041
GenBank mRNA AB066224.1 - 2 526
AF245000.1 - 709
D67087.1 - 2 499
L10152.1 - 2 493
U70476.1 - 2 578

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
basic amino acid transport
arginine transport
regulation of TOR signaling
cation transmembrane transport
basic amino acid transmembrane transporter activity
arginine transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0