This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A1 SIRNA Assays ›
Gene Symbol:
Slc7a1
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
Gene Aliases:
Atrc1, CAT1, Cat-1
Chromosome Location:
Chr.12: 8085012 - 8108972 on Build Rnor_6.0
UniGene ID:
Rn.9439
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303957
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013111.3 NP_037243.2 481
XM_008768943.1 XP_008767165.1 1 753
XM_008768944.1 XP_008767166.1 1 753
GenBank mRNA AB066224.1 - 1 238
AF245000.1 - 421
D67087.1 - 1 211
L10152.1 - 1 205
U70476.1 - 1 290

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
basic amino acid transport
arginine transport
regulation of TOR signaling
cation transmembrane transport
basic amino acid transmembrane transporter activity
arginine transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0