This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other F2R SIRNA Assays ›
Gene Symbol:
F2r
Gene Name:
coagulation factor II (thrombin) receptor
Gene Aliases:
Par1, TRGPC
Chromosome Location:
Chr.2: 26118760 - 26135340 on Build Rnor_6.0
UniGene ID:
Rn.2609
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303633
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F2r
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012950.2 NP_037082.2 2 562
GenBank mRNA BC085759.1 - 2 562 7106133
M81642.1 - 2 570

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010169, AK146194, AK085990, AK160032, BC031516, and others.

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
connective tissue replacement involved in inflammatory response wound healing
negative regulation of glomerular filtration
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
protein kinase C-activating G-protein coupled receptor signaling pathway
tyrosine phosphorylation of STAT protein
STAT protein import into nucleus
establishment of synaptic specificity at neuromuscular junction
blood coagulation
positive regulation of cell proliferation
negative regulation of cell proliferation
response to wounding
positive regulation of phosphatidylinositol 3-kinase signaling
obsolete death
platelet activation
regulation of blood coagulation
positive regulation of blood coagulation
positive regulation of cell migration
response to lipopolysaccharide
regulation of interleukin-1 beta production
positive regulation of collagen biosynthetic process
positive regulation of Rho protein signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
positive regulation of JAK-STAT cascade
homeostasis of number of cells within a tissue
release of sequestered calcium ion into cytosol
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of calcium ion transport
regulation of sensory perception of pain
positive regulation of ERK1 and ERK2 cascade
thrombin receptor signaling pathway
negative regulation of renin secretion into blood stream
positive regulation of interleukin-8 secretion
positive regulation of interleukin-6 secretion
G-protein alpha-subunit binding
G-protein coupled receptor activity
receptor binding
thrombin receptor activity
G-protein beta-subunit binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header